siRNA Details
| Gene Symbol: 5430425K12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017339.1 | - | 2 | 534 | ||
| DV053914.1 | - | 5 | 831 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5430425K12Rik| Gene Symbol: 5430425K12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017339.1 | - | 2 | 534 | ||
| DV053914.1 | - | 5 | 831 | |||