siRNA Details
Gene Symbol: Gm12657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081019.1 | NP_001074488.1 | 2 | 861 |
Gene Symbol: H3f3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008210.4 | NP_032236.1 | 4 | 972 | ||
GenBank mRNA | AI155695.1 | - | 1 | 49 | ||
AK142749.1 | - | 1 | 196 | |||
AK150928.1 | - | 4 | 927 | |||
AK152293.1 | - | 4 | 927 | |||
AK160677.1 | - | 4 | 931 | |||
AK168197.1 | - | 4 | 931 | |||
AK169608.1 | - | 4 | 927 | |||
BC002268.1 | - | 4 | 917 | 3495830 | ||
BC012687.1 | - | 4 | 921 | 4224280 | ||
BC083353.1 | - | 4 | 912 | 5714859 | ||
BC088835.1 | - | 4 | 909 | 6437687 | ||
BC106177.1 | - | 4 | 905 | 6511787 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC027275.1 | - | 2447 | Chr. - - [-] on Build |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003750348, XM_002729644, XM_002728043, FQ218544, FQ220271, and others.
Panther Classification:
Gene Ontology Categories:
