siRNA Details
| Gene Symbol: I830012O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005858.3 | NP_001005858.2 | 168 | |||
| GenBank mRNA | AK077243.1 | - | 157 | |||
| AK150632.1 | - | 2 | 415 | |||
| AK151442.1 | - | 2 | 415 | |||
| AK152620.1 | - | 2 | 415 | |||
| CA576017.1 | - | 168 | ||||

I830012O16Rik
Genomic Map