siRNA Details
| Gene Symbol: Gm14446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101605.1 | NP_001095075.1 | 2 | 1139 | ||
| NM_001110517.1 | NP_001103987.1 | 2 | 1220 | |||
| GenBank mRNA | AA175618.1 | - | 1 | 59 | ||
| AK090072.1 | - | 2 | 1216 | |||

Gm14446
Genomic Map