siRNA Details
| Gene Symbol: Gm14446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101605.1 | NP_001095075.1 | 2 | 962 | ||
| NM_001110517.1 | NP_001103987.1 | 2 | 1043 | |||
| GenBank mRNA | AK090072.1 | - | 2 | 1039 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14446| Gene Symbol: Gm14446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101605.1 | NP_001095075.1 | 2 | 962 | ||
| NM_001110517.1 | NP_001103987.1 | 2 | 1043 | |||
| GenBank mRNA | AK090072.1 | - | 2 | 1039 | ||