siRNA Details
Gene Symbol: Vmn1r142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166749.1 | NP_001160221.1 | 1 | 392 |
Gene Symbol: Vmn1r94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166723.1 | NP_001160195.1 | 1 | 392 |
