siRNA Details
| Gene Symbol: Zfp600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177545.2 | NP_001171016.1 | 5 | 2060 | ||
| NM_001177546.2 | NP_001171017.1 | 5 | 2153 | |||
| GenBank mRNA | BC108352.1 | - | 5 | 2060 | 30462028 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp600| Gene Symbol: Zfp600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177545.2 | NP_001171016.1 | 5 | 2060 | ||
| NM_001177546.2 | NP_001171017.1 | 5 | 2153 | |||
| GenBank mRNA | BC108352.1 | - | 5 | 2060 | 30462028 | |