siRNA Details
| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039485.4 | NP_001034574.4 | 34 | 5727 | ||
| GenBank mRNA | AK087529.1 | - | 33 | 4960 | ||
| HQ215521.1 | - | 34 | 4813 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Piezo2| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039485.4 | NP_001034574.4 | 34 | 5727 | ||
| GenBank mRNA | AK087529.1 | - | 33 | 4960 | ||
| HQ215521.1 | - | 34 | 4813 | |||