siRNA Details
| Gene Symbol: Tcra-V22.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U47051.1 | - | 2 | 202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcra-V22.1| Gene Symbol: Tcra-V22.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U47051.1 | - | 2 | 202 | ||