siRNA Details
| Gene Symbol: Zfp534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127188.2 | NP_001120660.1 | 4 | 789 | ||
| Gene Symbol: Gm13051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037926.2 | NP_001033015.1 | 4 | 744 | ||
| GenBank mRNA | BC108336.1 | - | 4 | 722 | 30462068 | |

Zfp534
Genomic Map