siRNA Details
| Gene Symbol: Rnf170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC031383.1 | - | 8 | 1214 | ||
| BC138930.1 | - | 8 | 1257 | 8861952 | ||
| BC145718.1 | - | 8 | 1262 | 40131021 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf170| Gene Symbol: Rnf170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC031383.1 | - | 8 | 1214 | ||
| BC138930.1 | - | 8 | 1257 | 8861952 | ||
| BC145718.1 | - | 8 | 1262 | 40131021 | ||