siRNA Details
| Gene Symbol: Syce1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048145.1 | NP_001041610.1 | 5 | 332 | ||
| NM_001286494.1 | NP_001273423.1 | 4 | 269 | |||
| NM_029139.1 | NP_083415.1 | 5 | 332 | |||
| GenBank mRNA | AK015606.1 | - | 5 | 328 | ||
| BB616021.1 | - | 5 | 312 | |||
| BC100478.1 | - | 4 | 250 | |||
| BC141326.1 | - | 5 | 332 | 9055938 | ||
| BC141347.1 | - | 5 | 332 | 9055959 | ||

Syce1l
Genomic Map