siRNA Details
| Gene Symbol: Zxdb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081473.1 | NP_001074942.1 | 1 | 2213 | ||
| GenBank mRNA | AK016708.1 | - | 1 | 1301 | ||
| AK132358.1 | - | 1 | 1356 | |||
| Gene Symbol: Zxda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK155431.1 | - | 2 | 1455 | ||

Zxdb
Genomic Map