siRNA Details
| Gene Symbol: Pex10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042407.1 | NP_001035866.1 | 2 | 175 | ||
| GenBank mRNA | AK043682.1 | - | 2 | 149 | ||
| BC147304.1 | - | 2 | 160 | 9056372 | ||
| BC147305.1 | - | 2 | 160 | 9056374 | ||
| CJ064027.1 | - | 2 | 149 | |||
| CJ176766.1 | - | 2 | 176 | |||

Pex10
Genomic Map