siRNA Details
| Gene Symbol: Pex10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042407.1 | NP_001035866.1 | 3 | 622 | ||
| GenBank mRNA | BC147304.1 | - | 3 | 607 | 9056372 | |
| BC147305.1 | - | 3 | 607 | 9056374 | ||
| CB203398.1 | - | 2 | 196 | |||
| CJ064027.1 | - | 3 | 596 | |||

Pex10
Genomic Map