siRNA Details
| Gene Symbol: Mettl7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081471.1 | NP_001074940.1 | 2 | 547 | ||
| GenBank mRNA | AB253945.1 | - | 2 | 547 | ||
| Gene Symbol: Mettl7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027334.3 | NP_081610.2 | 2 | 576 | ||
| GenBank mRNA | AB253943.1 | - | 2 | 547 | ||
| AJ536232.1 | - | 2 | 550 | |||
| AK076169.1 | - | 2 | 578 | |||
| BC033446.1 | - | 2 | 553 | |||
| BC047149.1 | - | 2 | 560 | |||
| BC055034.1 | - | 2 | 567 | 6306873 | ||
| BC060144.1 | - | 2 | 555 | |||
| BC171988.1 | - | 2 | 570 | 9054274 | ||
| BC171989.1 | - | 2 | 570 | 9054275 | ||
| Gene Symbol: Mettl7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199477.2 | NP_955771.2 | 2 | 556 | ||
| GenBank mRNA | AB099518.1 | - | 2 | 593 | ||
| AB253944.1 | - | 2 | 547 | |||
| AK143923.1 | - | 2 | 557 | |||
| BC087869.1 | - | 2 | 556 | 30304748 | ||
| BC090971.1 | - | 2 | 556 | 30314995 | ||

Mettl7a3
Genomic Map