siRNA Details
| Gene Symbol: Esp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244651.1 | NP_001231580.1 | 3 | 128 | ||
| GenBank mRNA | AB306993.1 | - | 3 | 128 | ||
| Gene Symbol: Esp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001255977.1 | NP_001242906.1 | 3 | 210 | ||
| GenBank mRNA | AB306995.1 | - | 3 | 210 | ||

Esp15
Genomic Map