siRNA Details
| Gene Symbol: LOC668306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG086570.2 | - | 149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC668306| Gene Symbol: LOC668306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG086570.2 | - | 149 | |||