siRNA Details
| Gene Symbol: CXCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002994.4 | NP_002985.1 | 2 | 294 | ||
| GenBank mRNA | BC008376.1 | - | 2 | 245 | 3826939 | |
| BT006807.1 | - | 2 | 146 | |||
| BU169770.1 | - | 2 | 265 | |||
| CR457428.1 | - | 2 | 146 | |||
| DN992282.1 | - | 2 | 352 | |||
| X78686.1 | - | 2 | 252 | |||

CXCL5
Genomic Map