siRNA Details
| Gene Symbol: Dmrtc1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142690.1 | NP_001136162.1 | 9 | 1494 | ||
| Gene Symbol: Dmrtc1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142691.1 | NP_001136163.1 | 9 | 1494 | ||
| GenBank mRNA | AK016891.1 | - | 9 | 1495 | ||

Dmrtc1c2
Genomic Map