siRNA Details
| Gene Symbol: Gm9125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163730.2 | NP_001157202.1 | 2 | 2026 | ||
| GenBank mRNA | AK033449.1 | - | 2 | 2027 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9125| Gene Symbol: Gm9125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163730.2 | NP_001157202.1 | 2 | 2026 | ||
| GenBank mRNA | AK033449.1 | - | 2 | 2027 | ||