siRNA Details
| Gene Symbol: Pgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008828.3 | NP_032854.2 | 4, 5 | 569 | ||
| GenBank mRNA | AK133877.1 | - | 4, 5 | 491 | ||
| AK145846.1 | - | 4, 5 | 505 | |||
| AK164440.1 | - | 4, 5 | 491 | |||
| AK167441.1 | - | 4, 5 | 463 | |||
| AK167459.1 | - | 4, 5 | 463 | |||
| AK167710.1 | - | 4, 5 | 462 | |||
| AK180821.1 | - | 2 | 223 | |||
| BC023819.1 | - | 4, 5 | 475 | |||
| BC032167.1 | - | 4, 5 | 475 | |||
| BC083355.1 | - | 4, 5 | 461 | 6828087 | ||
| BC108372.1 | - | 4, 5 | 473 | 5060103 | ||

Pgk1
Genomic Map