siRNA Details
| Gene Symbol: Zfp507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177739.3 | NP_808407.2 | 6 | 2646 | ||
| GenBank mRNA | AK129283.1 | - | 6 | 2614 | ||
| AK162510.1 | - | 4 | 826 | |||
| BC050041.1 | - | 6 | 2635 | 30006851 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp507| Gene Symbol: Zfp507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177739.3 | NP_808407.2 | 6 | 2646 | ||
| GenBank mRNA | AK129283.1 | - | 6 | 2614 | ||
| AK162510.1 | - | 4 | 826 | |||
| BC050041.1 | - | 6 | 2635 | 30006851 | ||