siRNA Details
| Gene Symbol: Zfp133-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137157.1 | - | 2 | 98 | ||
| DT931029.1 | - | 2 | 145 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp133-ps| Gene Symbol: Zfp133-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137157.1 | - | 2 | 98 | ||
| DT931029.1 | - | 2 | 145 | |||