siRNA Details
| Gene Symbol: Lpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008509.2 | NP_032535.2 | 10 | 1764 | ||
| GenBank mRNA | AK002645.1 | - | 10 | 1690 | ||
| AK045064.1 | - | 10 | 1653 | |||
| AK079446.1 | - | 10 | 1653 | |||
| AK086023.1 | - | 10 | 1585 | |||
| AK150328.1 | - | 10 | 1765 | |||
| AK150355.1 | - | 10 | 1765 | |||
| AK150375.1 | - | 10 | 1763 | |||
| AK150457.1 | - | 10 | 1660 | |||
| AK150488.1 | - | 10 | 1765 | |||
| AK150505.1 | - | 10 | 1765 | |||
| AK150532.1 | - | 10 | 1765 | |||
| AK150577.1 | - | 10 | 1765 | |||
| AK150585.1 | - | 10 | 1765 | |||
| AK150593.1 | - | 10 | 1765 | |||
| AK150646.1 | - | 10 | 1762 | |||
| AK150672.1 | - | 10 | 1683 | |||
| AK150691.1 | - | 10 | 1765 | |||
| AK150984.1 | - | 10 | 1765 | |||
| AK150990.1 | - | 10 | 1765 | |||
| AK151038.1 | - | 10 | 1663 | |||
| AK151049.1 | - | 10 | 1765 | |||
| AK151093.1 | - | 10 | 1765 | |||
| AK151099.1 | - | 10 | 1765 | |||
| AK151105.1 | - | 10 | 1765 | |||
| AK151112.1 | - | 10 | 1766 | |||
| AK151195.1 | - | 10 | 1663 | |||
| AK151216.1 | - | 10 | 1766 | |||
| AK151369.1 | - | 10 | 1683 | |||
| AK151434.1 | - | 10 | 1683 | |||
| AK151521.1 | - | 10 | 1683 | |||
| AK151563.1 | - | 10 | 1765 | |||
| AK151630.1 | - | 10 | 1663 | |||
| AK151679.1 | - | 10 | 1765 | |||
| AK151727.1 | - | 10 | 1765 | |||
| AK151772.1 | - | 10 | 1765 | |||
| AK151828.1 | - | 10 | 1653 | |||
| AK151866.1 | - | 10 | 1765 | |||
| AK151870.1 | - | 10 | 1765 | |||
| AK151872.1 | - | 10 | 1765 | |||
| AK152014.1 | - | 10 | 1765 | |||
| AK152035.1 | - | 10 | 1765 | |||
| AK152049.1 | - | 10 | 1765 | |||
| AK152053.1 | - | 10 | 1653 | |||
| AK152079.1 | - | 10 | 1765 | |||
| AK152238.1 | - | 10 | 1766 | |||
| AK152251.1 | - | 10 | 1763 | |||
| AK152350.1 | - | 10 | 1765 | |||
| AK152390.1 | - | 10 | 1604 | |||
| AK152481.1 | - | 10 | 1766 | |||
| AK152657.1 | - | 10 | 1765 | |||
| AK152706.1 | - | 10 | 1766 | |||
| AK152733.1 | - | 10 | 1663 | |||
| AK152775.1 | - | 10 | 1765 | |||
| AK152869.1 | - | 10 | 1765 | |||
| AK152956.1 | - | 10 | 1766 | |||
| AK153006.1 | - | 10 | 1765 | |||
| AK153148.1 | - | 10 | 1765 | |||
| AK153242.1 | - | 10 | 1766 | |||
| AK153425.1 | - | 10 | 1765 | |||
| AK153428.1 | - | 10 | 1765 | |||
| AK159268.1 | - | 10 | 1765 | |||
| AK170486.1 | - | 10 | 1764 | |||
| BC003305.1 | - | 10 | 1753 | 3584788 | ||
| BC158040.1 | - | 10 | 1598 | 9088156 | ||

Lpl
Genomic Map