siRNA Details
| Gene Symbol: Parp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163575.1 | NP_001157047.1 | 5 | 886 | ||
| NM_001163576.1 | NP_001157048.1 | 4 | 990 | |||
| GenBank mRNA | AK028373.1 | - | 5 | 834 | ||
| AK035309.1 | - | 4 | 960 | |||
| AK046755.1 | - | 4 | 971 | |||
| AK146336.1 | - | 5 | 887 | |||
| AK153205.1 | - | 4 | 727 | |||
| AK166291.1 | - | 5 | 837 | |||
| BC024074.1 | - | 5 | 843 | |||

Parp10
Genomic Map