siRNA Details
| Gene Symbol: Parp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163575.1 | NP_001157047.1 | 9, 10 | 2497 | ||
| NM_001163576.1 | NP_001157048.1 | 8, 9 | 2601 | |||
| GenBank mRNA | AK028373.1 | - | 9, 10 | 2445 | ||
| AK035309.1 | - | 8, 9 | 2621 | |||
| AK046755.1 | - | 8, 9 | 2582 | |||
| AK146336.1 | - | 9, 10 | 2497 | |||
| AK153205.1 | - | 8, 9 | 2338 | |||
| AK166291.1 | - | 9, 10 | 2448 | |||
| BC023056.1 | - | 5, 6 | 1334 | |||
| BC024074.1 | - | 9, 10 | 2454 | |||
| BC025608.1 | - | 5, 6 | 1479 | |||

Parp10
Genomic Map