siRNA Details
| Gene Symbol: Sms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009214.3 | NP_033240.3 | 4 | 396 | ||
| GenBank mRNA | AF031486.1 | - | 4 | 379 | ||
| AK017775.1 | - | 4 | 367 | |||
| AK049780.1 | - | 4 | 395 | |||
| AK049787.1 | - | 4 | 385 | |||
| AK136798.1 | - | 4 | 397 | |||
| AK149929.1 | - | 4 | 394 | |||
| AK160659.1 | - | 4 | 397 | |||
| AK166665.1 | - | 4 | 422 | |||
| BC046623.1 | - | 4 | 378 | 6315420 | ||
| BC058688.1 | - | 4 | 637 | 6413869 | ||
| Y09419.1 | - | 4 | 369 | |||

Sms
Genomic Map