siRNA Details
| Gene Symbol: E330020D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143622.1 | - | 2 | 977 | ||
| AK168480.1 | - | 2 | 975 | |||
| Gene Symbol: LOC432459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082906.1 | - | 1 | 1853 | ||

E330020D12Rik
Genomic Map