siRNA Details
Gene Symbol: Gm14625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220498.1 | NP_001207427.1 | 3 | 292 | ||
GenBank mRNA | BC145074.1 | - | 292 | 9053622 | ||
BC171942.1 | - | 3 | 292 | 9054228 |
Gene Symbol: Gm16404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220497.1 | NP_001207426.1 | 3 | 292 | ||
GenBank mRNA | BC171941.1 | - | 3 | 292 | 9054227 |
Gene Symbol: Gm16405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166646.1 | NP_001160118.1 | 294 |
Gene Symbol: Gm16430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166601.1 | NP_001160073.1 | 3 | 294 | ||
GenBank mRNA | BC108390.1 | - | 3 | 294 |
Gene Symbol: Slxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029181.1 | NP_083457.1 | 3 | 329 | ||
GenBank mRNA | AK015913.1 | - | 3 | 330 | ||
BC108389.1 | - | 297 |
