siRNA Details
| Gene Symbol: Gbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194336.2 | NP_919317.2 | 11 | 1777 | ||
| GenBank mRNA | AK171913.1 | - | 8 | 3415 | ||
| BC007143.1 | - | 10 | 1634 | 3495728 | ||
| BC052867.1 | - | 6 | 1005 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbp6| Gene Symbol: Gbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194336.2 | NP_919317.2 | 11 | 1777 | ||
| GenBank mRNA | AK171913.1 | - | 8 | 3415 | ||
| BC007143.1 | - | 10 | 1634 | 3495728 | ||
| BC052867.1 | - | 6 | 1005 | |||