siRNA Details
| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144868.3 | NP_659117.2 | 6 | 2027 | ||
| GenBank mRNA | AK171516.1 | - | 5 | 2266 | ||
| BC038056.1 | - | 1 | 135 | |||
| BC054751.1 | - | 6 | 1306 | |||
| BC055289.1 | - | 7 | 1830 | |||
| BC096538.1 | - | 6 | 2027 | |||

Pcnxl3
Genomic Map