siRNA Details
| Gene Symbol: Ptpdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301781.1 | NP_001288710.1 | 10, 11 | 2626 | ||
| NM_207232.2 | NP_997115.1 | 9, 10 | 2443 | |||
| GenBank mRNA | AB185283.1 | - | 2, 3 | 227 | ||
| AK030415.1 | - | 9, 10 | 2436 | |||
| AK038922.1 | - | 9, 10 | 2356 | |||
| AK132781.1 | - | 10, 11 | 2535 | |||
| BC066081.1 | - | 9, 10 | 2344 | 30355705 | ||
| BC068267.1 | - | 2, 3 | 288 | |||

Ptpdc1
Genomic Map