siRNA Details
| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005065.5 | NP_005056.3 | 19, 20 | 2171 | ||
| XM_005267988.2 | XP_005268045.1 | 19, 20 | 2054 | |||
| XM_005267989.3 | XP_005268046.1 | 18, 19 | 2105 | |||
| GenBank mRNA | AB020335.1 | - | 19, 20 | 2099 | ||
| AF052059.1 | - | 19, 20 | 2147 | |||
| BC040498.1 | - | 2, 3 | 258 | |||
| U11037.1 | - | 1, 2 | 151 | |||

SEL1L
Genomic Map