siRNA Details
| Gene Symbol: St6galnac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC141095.1 | - | 1558 | 9055707 | ||
| BC145373.1 | - | 1558 | 9053931 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St6galnac1| Gene Symbol: St6galnac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC141095.1 | - | 1558 | 9055707 | ||
| BC145373.1 | - | 1558 | 9053931 | |||