siRNA Details
| Gene Symbol: SEPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085486.1 | NP_001078955.1 | 3, 4 | 329 | ||
| NM_001093726.1 | NP_001087195.1 | 3, 4 | 388 | |||
| NM_005410.2 | NP_005401.3 | 2, 3 | 300 | |||
| GenBank mRNA | AK096125.1 | - | 2, 3 | 669 | ||
| AK225801.1 | - | 3, 4 | 299 | |||
| AK311392.1 | - | 2, 3 | 637 | |||
| AL833145.1 | - | 2, 3 | 271 | |||
| AU136519.2 | - | 3, 4 | 358 | |||
| BC005244.1 | - | 2, 3 | 270 | |||
| BC015875.1 | - | 2, 3 | 274 | 4715833 | ||
| BC040075.1 | - | 2, 3 | 250 | 5770225 | ||
| BC046152.1 | - | 2, 3 | 251 | 6192976 | ||
| BC058919.1 | - | 2, 3 | 281 | 6195379 | ||
| BI463468.1 | - | 2, 3 | 445 | |||

SEPP1
Genomic Map