siRNA Details
Gene Symbol: Slc30a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC013478.1 | - | 3 | 741 | ||
BC147704.1 | - | 4 | 552 | 9087463 | ||
BC147707.1 | - | 4 | 552 | 9087466 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc30a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC013478.1 | - | 3 | 741 | ||
BC147704.1 | - | 4 | 552 | 9087463 | ||
BC147707.1 | - | 4 | 552 | 9087466 |