siRNA Details
| Gene Symbol: Vmn2r76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102580.1 | NP_001096050.1 | 5 | 1556 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r76| Gene Symbol: Vmn2r76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102580.1 | NP_001096050.1 | 5 | 1556 | ||
Set Membership: |
|