siRNA Details
| Gene Symbol: Psma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011967.3 | NP_036097.1 | 4 | 599 | ||
| GenBank mRNA | AF019661.1 | - | 4 | 314 | ||
| AK143468.1 | - | 4 | 601 | |||
| AK160530.1 | - | 4 | 359 | |||
| AK160550.1 | - | 4 | 353 | |||
| AK160741.1 | - | 4 | 362 | |||
| AK161680.1 | - | 4 | 347 | |||
| BC007181.1 | - | 4 | 346 | |||
| BC010709.1 | - | 4 | 335 | 2648635 | ||
| BC083342.1 | - | 4 | 332 | 5700963 | ||
| BC108368.1 | - | 4 | 329 | 3596530 | ||

Psma5
Genomic Map