siRNA Details
| Gene Symbol: Vmn1r224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166735.1 | NP_001160207.1 | 1 | 219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r224| Gene Symbol: Vmn1r224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166735.1 | NP_001160207.1 | 1 | 219 | ||