siRNA Details
| Gene Symbol: 3110040M04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014162.1 | - | 1 | 1457 | ||
| Gene Symbol: Gm8947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AV000681.1 | - | 268 | |||

3110040M04Rik
Genomic Map