siRNA Details
| Gene Symbol: Immp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028260.2 | NP_082536.1 | 6 | 566 | ||
| GenBank mRNA | AK005356.1 | - | 6 | 567 | ||
| AK009570.1 | - | 6 | 524 | |||
| AK010075.1 | - | 6 | 524 | |||
| AK011278.1 | - | 6 | 567 | |||
| AK011382.1 | - | 6 | 567 | |||
| AK011420.1 | - | 6 | 528 | |||
| AK011466.1 | - | 6 | 539 | |||
| AK011618.1 | - | 6 | 528 | |||
| AK011872.1 | - | 6 | 530 | |||
| AK012181.1 | - | 519 | ||||
| AK015978.1 | - | 6 | 537 | |||
| AK078300.1 | - | 6 | 553 | |||
| AK135670.1 | - | 6 | 526 | |||
| AK168202.1 | - | 6 | 515 | |||
| BC008259.1 | - | 6 | 525 | 2649406 | ||
| BC081433.1 | - | 5 | 409 | 6814816 | ||

Immp1l
Genomic Map