siRNA Details
Gene Symbol: Wnt8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011720.3 | NP_035850.2 | 6 | 2093 | ||
GenBank mRNA | AK169459.1 | - | 2 | 1593 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wnt8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011720.3 | NP_035850.2 | 6 | 2093 | ||
GenBank mRNA | AK169459.1 | - | 2 | 1593 |