siRNA Details
| Gene Symbol: Wnt8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011720.3 | NP_035850.2 | 6 | 2063 | ||
| GenBank mRNA | AK169459.1 | - | 2 | 1563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt8b| Gene Symbol: Wnt8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011720.3 | NP_035850.2 | 6 | 2063 | ||
| GenBank mRNA | AK169459.1 | - | 2 | 1563 | ||