siRNA Details
| Gene Symbol: Zfp146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011980.3 | NP_036110.1 | 2 | 948 | ||
| GenBank mRNA | AJ224763.1 | - | 2 | 911 | ||
| AK047696.1 | - | 2 | 956 | |||
| BC127038.1 | - | 1 | 389 | 40051938 | ||
| BC127039.1 | - | 1 | 390 | 40051941 | ||

Zfp146
Genomic Map