siRNA Details
| Gene Symbol: Olfr208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146384.1 | NP_666496.1 | 1 | 103 | ||
| Gene Symbol: Olfr207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011792.1 | NP_001011792.1 | 1 | 103 | ||
| Gene Symbol: Olfr209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207551.2 | NP_997434.2 | 1 | 103 | ||

Olfr208
Genomic Map