siRNA Details
| Gene Symbol: Wfdc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081957.1 | NP_001075426.1 | 2 | 227 | ||
| GenBank mRNA | BC038870.1 | - | 2 | 156 | ||
| BC089618.1 | - | 2 | 200 | 6766535 | ||
| FJ007372.1 | - | 2 | 268 | |||
| W08116.1 | - | 2 | 227 | |||

Wfdc17
Genomic Map