siRNA Details
| Gene Symbol: SRSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005626.4 | NP_005617.2 | 2 | 558 | ||
| XM_011541951.2 | XP_011540253.1 | 2 | 358 | |||
| GenBank mRNA | AB209856.1 | - | 5 | 5498 | ||
| AK223444.1 | - | 2 | 335 | |||
| AK291509.1 | - | 2 | 280 | |||
| AK293706.1 | - | 2 | 331 | |||
| AK310528.1 | - | 5 | 818 | |||
| BC002781.2 | - | 2 | 282 | 3619538 | ||
| BT007415.1 | - | 2 | 185 | |||
| L14076.1 | - | 2 | 232 | |||

SRSF4
Genomic Map