siRNA Details
| Gene Symbol: SRSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005626.4 | NP_005617.2 | 5 | 1020 | ||
| GenBank mRNA | AB209856.1 | - | 7 | 6850 | ||
| AK223444.1 | - | 5 | 797 | |||
| AK291509.1 | - | 5 | 742 | |||
| BC002781.2 | - | 5 | 744 | 3619538 | ||
| BT007415.1 | - | 5 | 647 | |||
| L14076.1 | - | 5 | 694 | |||

SRSF4
Genomic Map