siRNA Details
| Gene Symbol: Acly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111095.1 | NP_001104565.1 | 17 | 2093 | ||
| NM_016987.2 | NP_058683.2 | 18 | 2123 | |||
| XM_006247254.2 | XP_006247316.1 | 18 | 2116 | |||
| GenBank mRNA | BC100618.1 | - | 17 | 2085 | 7934668 | |
| J05210.1 | - | 18 | 2071 | |||

Acly
Genomic Map